Cell scrapers and cell lifters, Corning®
Supplier: Corning
Certificates
About this item
Useful for the manual harvesting of cells, scrapers are designed for use in flasks and cell lifters for use in harvesting cells (especially stem cells) in dishes.
- Blade design minimises cell damage and ensures even contact with the growth surface
- Individually wrapped
- Sterilised by gamma radiation
- Certified non-pyrogenic
Specifications
- Catalog No:
- 734-1526
- 734-1528
- 734-1527
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa