ContactUsLinkComponent
Storage plates and blocks, 96-well, Corning®
Storage plates and blocks, 96-well, Corning®
Catalog # CORN3947
Supplier:  Corning
undefined
Storage plates and blocks, 96-well, Corning®
Catalog # CORN3947
Supplier:  Corning
Specifications
  • Packed:
    5/bag
  • Sterile:
    -
  • Description:
    Deep well plate, 96-well, natural, V-bottom square well
  • Seq:
    03
  • W×D×H:
    127,76×85,48×44,2 mm
  • Pk:
    50
  • Well volume:
    2200 µl
Specifications
About this item
PP, non treated, without lid. Standard and large volume 96-well microplates and storage blocks to meet assay and storage requirements.
  • Choice of flat, round or V-shaped well bottom
  • Uniform skirt heights for greater robotic gripping surface
  • Resistant to many common organic solvents (e.g. DMSO, ethanol, methanol)
  • Certified free from DNase and RNase