Green long-term cell tracer, CYTO-ID®
Catalog # ENZOENZ51036K025
Lieferant: Enzo Life Sciences
CAS-Nummer:
Spezifikationen
- Pk:1 KIT
- Description:Green long-term cell tracer for 25 assays
- Seq:02
Spezifikationen
Über diesen Artikel
Live cell fluorescent labelling used for a long term cell viability, cytotoxicity, cell adhesion, cell migration, and cell-cell fusion studies.
- Dual labelling with CELLESTIAL® fluorescent probes
- Minimal transfer of fluorescence from dye-labeled to unlabeled cells
- Proprietary non covalent cell labeling technology to stably incorporate a green fluorescent dye containing hydrophobic aliphatic chains into the cell membrane’s lipid bilayer
Ordering information: Contains a green tracer dye (50 µl), 4X labeling buffer (12,5 ml), and 10X HBSS (25 ml).