Autoclave Bags, standard
Supplier: RATIOLAB
Certificates
About this item
Bags of very strong PP-film with a broad seam at the bottom, for the disposal of contaminated labware.
- Autoclavable at 134 °C (20 min.)
- Film thickness 50 or 100 µm
For autoclaving, the bag must not be completely closed.
In dispenser box
Product Information
Frequently Bought Together