Fingertight capillary column nuts
Supplier: Restek
Certificates
About this item
These fingertight capillary column nuts rapidly tighten without wrenches and can be used with 0,25, 0,32, or 0,53 mm ID columns.
Frequently Bought Together
These fingertight capillary column nuts rapidly tighten without wrenches and can be used with 0,25, 0,32, or 0,53 mm ID columns.