Air diverter for Agilent GCs
Catalog # REST22076
Supplier: Restek
Specifications
- Description:Air diverter for Agilent 5890/6890/7890 GCs
- Pk:1
Specifications
About this item
This air diverter diverts GC exhaust heat up and away from the lab bench.
- Easy to install and no tools required
- Compatible with 10,16 cm flexible metal dryer hose