ContactUsLinkComponent
Direct injection liner adapter
Direct injection liner adapter
Catalog # REST21303
Supplier:  Restek
Direct injection liner adapter
Catalog # REST21303
Supplier:  Restek
Specifications
  • Description:
    Direct injection liner adapter for Agilent 5890 packed column GC
  • Pk:
    1
Specifications
About this item
This direct injection liner adapter uses standard 1,59 mm capillary nut and ferrules. Can be used with Agilent or Restek direct injection glass inserts or Restek direct injection uniliner liners for an Agilent 5890 packed column GC.
  • Convenient wrench pad at base
Delivery: Adapter includes 6,35 mm graphite ferrule and stainless steel nut.