Orders LinkContactUsLinkComponent
Agarose, SeaKem® HEEO
Agarose, SeaKem® HEEO
Supplier:   LONZA
Agarose, SeaKem® HEEO
Supplier:  LONZA
Certificates
Agarose, SeaKem® HEEO
Agarose, SeaKem® HEEO
2 Options Available Below
Product Details & Documents
SeaKem® HEEO Agarose is a very high EEO agarose that is useful in applications requiring significant cathodal migration, such as IEP of IgG and IgM. It can also be used to blend with a lower EEO agarose to achieve a specific EEO value.
Documents
Product Family Options
Product Information
  • Availability
    Price
  • 733-1558
    Agarose, SeaKem® HEEO
    Agarose, SeaKem® HEEO
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 220,00 €
    220,00 €
    1 * 25 g
       220,00 €
  • 733-1557
    Agarose, SeaKem® HEEO
    Agarose, SeaKem® HEEO
    Log in for availability
    Log in for availability
    1 * 125 g
    1 * 125 g - 727,00 €
    727,00 €
    1 * 125 g
       727,00 €