ContactUsLinkComponent
Examination gloves, nitrile, Sempercare® nitrile shine+
Examination gloves, nitrile, Sempercare® nitrile shine+
Certificates
Examination gloves, nitrile, Sempercare® nitrile shine+
Specifications
  • Catalog No:
  • SEMP826778433
  • SEMP826778435
  • SEMP826778437
  • Ambidextrous / hand-specific:
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Disposable / reusable:
  • Disposable
  • Disposable
  • Disposable
  • Latex / latex-free:
  • Latex-free
  • Latex-free
  • Latex-free
  • Powdered / powder-free:
  • Powder-free
  • Powder-free
  • Powder-free
  • Finish:
  • Textured fingertips
  • Textured fingertips
  • Textured fingertips
  • Cuff style:
  • Beaded
  • Beaded
  • Beaded
  • EN norm:
  • EN 455, EN 374-1/2-/3
  • EN 455, EN 374-1/2-/3
  • EN 455, EN 374-1/2-/3