ContactUsLinkComponent
Quartz Extraction Thimble for Emission Testing, Tapered, Whatman™
Quartz Extraction Thimble for Emission Testing, Tapered, Whatman™
Catalog # 537-0028
undefined
Quartz Extraction Thimble for Emission Testing, Tapered, Whatman™
Catalog # 537-0028
Specifications
  • Int.Ø×H:
    25×90 mm
  • Type:
    Tapered
  • Thickness:
    1,9 mm
  • Pk:
    10
Specifications
About this item
High-purity Whatman quartz thimbles from Cytiva support accurate, reproducible emission testing by providing low extractable levels and temperature resistance up to 800 °C.
  • Appropriate for manual sampling in aggressive environments
  • Suitable for both solvent extraction and air sampling applications
  • High-purity quartz/silica, for high-temperature resistance (up to 800 °C)
  • No binders or chemical additives
  • Low heavy metal content
  • High collection efficiency
Each Whatman quartz thimble from Cytiva is made from high-purity quartz with low heavy metal concentrations. Thimbles are 90 mm in length, tapered for stack sample loading, and have an outside diameter of 25 mm.