Wipes for general applications, VeraClean® Critical Cleaning Plus
Supplier: PGI NONWOVEN
Certificates
About this item
These semi-reusable wipes are perfectly suited for precision cleaning and critical wiping tasks across a range of industries. Thanks to their composite construction they can instantly absorb a wide range of fluids and provide a dry at once effect.
- Durable, strong enough for multiple uses
- High absorbency: Ideal for easy removal of fluids and spills
- Extremely strong both wet and dry: Won't fall apart like paper products, even when used in combination with detergents, water or solvent
- Low linting