Orders LinkContactUsLinkComponent
Feeder cabinets
Feeder cabinets
Supplier:   Palbam
Feeder cabinets
Supplier:  Palbam
Certificates
Feeder cabinets
2 Options Available Below
Product Details & Documents
The feeder cabinets support the gowning operation. Made of stainless steel 304 with electropolished finish.
  • Front access doors are used for reloading and for storage purposes
  • The bottom cabinet can be used for storage or disposal
  • Perspex dispenser slots ensure that garments/disposables can be extracted one at a time and do not spill out
Delivery information: Typically available from stock for direct shipment from the supplier. Delivered as shipped assembled or flat packed.
Product Family Options
Product Information
  • W×D×H
    Availability
    Price
  • PALBFFCS-0540-SST
    Feeder cabinets, front load and front dispense with 4 compartments
    Feeder cabinets, front load and front dispense with 4 compartments
    500×400×2000 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 8 660,00 €
    8 660,00 €
    1 * 1 ST
       8 660,00 €
  • PALBFFCS-1040-SST
    Feeder cabinets, front load and front dispense with 8 compartments
    Feeder cabinets, front load and front dispense with 8 compartments
    1000×400×2000 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 16 450,00 €
    16 450,00 €
    1 * 1 ST
       16 450,00 €