Order Entry
Austria
Orders LinkContactUsLinkComponent
VWR®, Loading dye base

VWR®, Loading dye base

Loading dye base 50X, 25 mL
Catalog # 1B1812-25ML
Supplier:   VWR Chemicals
VWR®, Loading dye base
Catalog # 1B1812-25ML
Supplier:  VWR Chemicals
CAS Number:  
VWR®, Loading dye base
VWR®, Loading dye base

Loading dye base 50X, 25 mL

CAS Number:  
Price
87,00 €
Log in for availability
1 * 25 mL
Product Details & Documents

This loading dye base is used for custom loading buffer preparation. It is a concentrated solution containing patent blue, bromophenol blue and amaranth tracking dyes, which migrate at approximately 4 000, 400 and 10 bp, on a 1% agarose gel, respectively.

Eliminates the need of weigh messy dye powders.

Documents

Specifications

Specifications

  • Pk:
    25 mL
  • Description:
    Loading dye base 50X

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.