Thermal transfer printer labels, type B-499
Supplier: Brady
Jump to:
About this item
Printable, nylon cloth labels for thermal transfer printers.
- Die-cut labels are supplied on a roll
- Contains some adhesive backed wire marking materials, and also labels that are pre-printed with headers or templates
- Supplied with the ribbon seperate, not in the same enclosure as the tape/sleeves
This material is UL recognised with its respective ribbon. See the recommended ribbon series for the appropriate ribbon to use.
24 Options Available Below
Product Details & Documents
Printable, nylon cloth labels for thermal transfer printers.
- Die-cut labels are supplied on a roll
- Contains some adhesive backed wire marking materials, and also labels that are pre-printed with headers or templates
- Supplied with the ribbon seperate, not in the same enclosure as the tape/sleeves
This material is UL recognised with its respective ribbon. See the recommended ribbon series for the appropriate ribbon to use.
Product Family Options
Product Information
- TypeModelMaterialAdhesivePrinting methodColourPrinter modelWidthDimensionsAvailabilityPrice
- BRDY116382Printer labelsPrinter labelsB-499THT-151-499-1.5-SCNylonPermanent acrylicThermal transferWhiteBBP1250,80 mm6×50 mmLog in for availability1 * 1 Rolle192,00 €
- BRDY062375Printer labelsPrinter labelsB-499THT-143-499-3NylonPermanent acrylicThermal transferWhite360X-Plus II9 mmLog in for availability1 * 1 Rolle217,00 €
- BRDY217066Printer labelsPrinter labelsB-499BPT-602-499NylonPermanent acrylicThermal transferWhiteBBP129 mmLog in for availability1 * 1 Rolle212,00 €
- BRDY217067Printer labelsPrinter labelsB-499BPT-602-499YLNylonPermanent acrylicThermal transferYellowBBP129 mmLog in for availability1 * 1 Rolle243,00 €
- BRDY116379Printer labelsPrinter labelsB-499THT-152-499-1.5-SCNylonPermanent acrylicThermal transferWhiteBBP1225,40 mm9×25 mmLog in for availability1 * 1 Rolle192,00 €
- BRDY062376Printer labelsPrinter labelsB-499THT-144-499-3NylonPermanent acrylicThermal transferWhite360X-Plus II12 mmLog in for availability1 * 1 Rolle327,00 €
- BRDY217068Printer labelsPrinter labelsB-499BPT-603-499NylonPermanent acrylicThermal transferWhiteBBP1212 mmLog in for availability1 * 1 Rolle232,00 €
- BRDY217069Printer labelsPrinter labelsB-499BPT-603-499YLNylonPermanent acrylicThermal transferYellowBBP1212 mmLog in for availability1 * 1 Rolle265,00 €
- BRDY030542Printer labelsPrinter labelsB-499THT-68-499-10NylonPermanent acrylicThermal transferWhite360X-Plus II12×19 mmLog in for availability1 * 1 Rolle480,00 €
- BRDY116368Printer labelsPrinter labelsB-499THT-125-499-1.5-SCNylonPermanent acrylicThermal transferWhiteBBP1222,86 mm12×22 mmLog in for availability1 * 1.500 ST192,00 €
- BRDY217075Printer labelsPrinter labelsB-499BPT-611-499NylonPermanent acrylicThermal transferWhiteBBP1212×25 mmLog in for availability1 * 1 Rolle353,00 €
- BRDY217082Printer labelsPrinter labelsB-499BPT-620-499NylonPermanent acrylicThermal transferWhiteBBP1212×25 mmLog in for availability1 * 1 Rolle199,00 €
- BRDY116384Printer labelsPrinter labelsB-499THT-144-499-1.5-SCNylonPermanent acrylicThermal transferWhiteBBP1248,26 mm12×48 mmLog in for availability1 * 1.500 ST224,00 €
- BRDY116376Printer labelsPrinter labelsB-499THT-136-499-1.5-SCNylonPermanent acrylicThermal transferWhiteBBP1238,10 mm19×38 mmLog in for availability1 * 1 Rolle192,00 €
- BRDY030543Printer labelsPrinter labelsB-499THT-69-499-3NylonPermanent acrylicThermal transferWhite360X-Plus II20×36 mmLog in for availability1 * 1 Rolle349,00 €
- BRDY062358Printer labelsPrinter labelsB-499THT-126-499-3NylonPermanent acrylicThermal transferWhite360X-Plus II22×12 mmLog in for availability1 * 1 Rolle312,00 €
- BRDY217081Printer labelsPrinter labelsB-499BPT-618-499NylonPermanent acrylicThermal transferWhiteBBP1222×38 mmLog in for availability1 * 1 Rolle618,00 €
- BRDY019260Printer labelsPrinter labelsB-499THT-163-499-3NylonPermanent acrylicThermal transferWhite360X-Plus II25×9 mmLog in for availability1 * 1 Rolle312,00 €
- BRDY062378Printer labelsPrinter labelsB-499THT-152-499-3NylonPermanent acrylicThermal transferWhite360X-Plus II25×9 mmLog in for availability1 * 1 Rolle312,00 €
- BRDY804776Printer labelsPrinter labelsB-499BPT-659-499NylonPermanent acrylicThermal transferWhiteBBP1225×12 mmLog in for availability1 * 1 Rolle338,00 €
- BRDY217084Printer labelsPrinter labelsB-499BPT-621-499NylonPermanent acrylicThermal transferWhiteBBP1225×36 mmLog in for availability1 * 1 Rolle618,00 €
- BRDY116370Printer labelsPrinter labelsB-499THT-137-499-1.5-SCNylonPermanent acrylicThermal transferWhiteBBP1250,80 mm25×50 mmLog in for availability1 * 1.500 ST248,00 €
- BRDY217073Printer labelsPrinter labelsB-499BPT-608-499NylonPermanent acrylicThermal transferWhiteBBP1241×15 mmLog in for availability1 * 1 Rolle298,00 €
- BRDY062377Printer labelsPrinter labelsB-499THT-151-499-3NylonPermanent acrylicThermal transferWhite360X-Plus II50×6 mmLog in for availability1 * 1 Rolle312,00 €