FID flow measuring adaptor
REST21000
: Restek
- Description:FID flow measuring adaptor
- Pk:1
This adaptor is suitable for for Agilent 5890/6850/6890/7890 GCs.
- Makes setting flows easy
Meets or exceeds original manufacturers performance.
This adaptor is suitable for for Agilent 5890/6850/6890/7890 GCs.
Meets or exceeds original manufacturers performance.