ContactUsLinkComponent
Mega 108 Storage Cabinet for HPLC Columns
  TRNT50001
 :  TRIPP
Mega 108 Storage Cabinet for HPLC Columns
  TRNT50001
 :  TRIPP
  • Description:
    HPLC storage cabinet, with clear lids
  • W×D×H:
    381×406×406 mm
  • Pk:
    1
  • Weight:
    13 kg
HPLC storage cabinet with constructed with white PVC and acrylic doors.
  • Mega organiser cabinet with seven removable, slide-out trays
  • Cabinet has a total of 108 adjustable compartments
  • Removable dividers fit most analytical HPLC columns
  • Each tray is covered with an acrylic lid that is completely removable
 : Doors with different colours are available on request.