ContactUsLinkComponent
Wafer tweezers, perforated
Wafer tweezers, perforated
Catalog # OUTI39S-4
Supplier:  OUTILS RUBIS
undefined
Wafer tweezers, perforated
Catalog # OUTI39S-4
Supplier:  OUTILS RUBIS
Specifications
  • Type:
    39S-4
  • Description:
    With 4 teeth, perforated lower paddle
  • Length:
    128 mm
  • Seq:
    10
  • Pk:
    1
Specifications
About this item
Stainless steel. These tweezers are specially designed to handle different fragile and delicate materials of gallium or silicon wafers.
  • Perforated lower paddle allows liquids to drop off
  • Smooth polished non glare surface on the inside of the paddel in order to prevent chipping and marring the wafer
  • Satin finish on the outside of the paddle for glare prevention