ContactUsLinkComponent
Digital colony counter, Quebec®
Digital colony counter, Quebec®
Catalog # REIC13332800
Supplier:  REICHERT
undefined
Digital colony counter, Quebec®
Catalog # REIC13332800
Supplier:  REICHERT
Specifications
  • Description:
    Digital colony counter
  • Environmentally Preferable:
  • Electrical:
    220 V/50 Hz
  • Illumination:
    Darkfield
  • Magnification:
    1,5×
  • Pk:
    1
Specifications
About this item
Digital colony counter delivers easy and accurate count of bacteria colonies with a convenient, built-in digital counter that records and displays the number of counted colonies.
  • Glare free illumination for sharp contrast
  • Light spreads uniformly on the counting plate for easily distinguishing colonies
  • Adjustable culture dish holder for accommodating small culture dishes and rod for easy focusing
  • Lens rotates for easy loading of culture dishes
  • Built in tilt leg for height and angle adjustment