Orders LinkContactUsLinkComponent
Multi-packs, Proline® Plus
   Sustainable Options Available
Supplier:   Sartorius
Multi-packs, Proline® Plus
Supplier:  Sartorius
   Sustainable Options Available
Certificates
Multi-packs, Proline® Plus
8 Options Available Below
Product Details & Documents
These Multipacks combine a set of Proline® Plus mechanical pipettes with matching tip trays and useful accessories. They offer an opportunity to test and get started with Proline® Plus mechanical pipettes.
  • Ergonomic design with light pipetting forces, comfortable handle and finger support for reduced risk of strain injuries
  • Minimised risk of contamination with Safe-Cone Filters and full autoclavability
  • Highly durable with strengthened structure also for heavier use
It is the widest range of pipettes for various users and applications.
 : All packs include a calibration tool that also acts as a tube opener, as well as the required instructions.
Specifications for Product Family
Specifications
  • Catalog No:
  • 613-5732
  • 613-5733
  • 613-5739
  • 613-5738
  • 613-5734
  • 613-5735
  • 613-5736
  • 613-5737
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • Renewable energy certificate; ISO 14001
  • Renewable energy certificate; ISO 14001
  • Renewable energy certificate; ISO 14001
  • Renewable energy certificate; ISO 14001
  • Renewable energy certificate; ISO 14001
  • Renewable energy certificate; ISO 14001
  • Renewable energy certificate; ISO 14001
  • Renewable energy certificate; ISO 14001
Specifications
Product Family Options
Product Information
  • Availability
    Price
  • 613-5732
    3-pack 10, incl. three single channel pipettes (0,5 - 10 µl, 10 - 100 µl, 100 - 1000 µl), Optifit Tip tray boxes (1× 0,1 - 10 µl, 1× 0,5 - 200 µl, 1× 10 - 1000 µl) and a linear stand
    3-pack 10, incl. three single channel pipettes (0,5 - 10 µl, 10 - 100 µl, 100 - 1000 µl), Optifit Tip tray boxes (1× 0,1 - 10 µl, 1× 0,5 - 200 µl, 1× 10 - 1000 µl) and a linear stand
    Log in for availability
    Log in for availability
    1 * 1 Pack.
    1 * 1 Pack. - 725,00 €
    725,00 €
    1 * 1 Pack.
       725,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-5732
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact
  • 613-5733
    3-pack 20, incl. three single channel pipettes (2 - 20 µl, 20 - 200 µl, 100 - 1000 µl), Optifit Tip tray boxes (2× 0,5 - 200 µl, 1× 10 - 1000 µl) and a linear stand
    3-pack 20, incl. three single channel pipettes (2 - 20 µl, 20 - 200 µl, 100 - 1000 µl), Optifit Tip tray boxes (2× 0,5 - 200 µl, 1× 10 - 1000 µl) and a linear stand
    Log in for availability
    Log in for availability
    1 * 1 Pack.
    1 * 1 Pack. - 725,00 €
    725,00 €
    1 * 1 Pack.
       725,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-5733
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact
  • 613-5739
    3-pack MAX, incl. three single channel pipettes (100 - 1000 µl, 500 - 5000 µl, 1000 - 10000 µl), Optifit Tip tray boxes (1× 10 - 1000 µl), Optifit Bulk-in-Bag tip packs (1× 5 ml, 1× 10 ml) and a linear stand
    3-pack MAX, incl. three single channel pipettes (100 - 1000 µl, 500 - 5000 µl, 1000 - 10000 µl), Optifit Tip tray boxes (1× 10 - 1000 µl), Optifit Bulk-in-Bag tip packs (1× 5 ml, 1× 10 ml) and a linear stand
    Log in for availability
    Log in for availability
    1 * 1 Pack.
    1 * 1 Pack. - 725,00 €
    725,00 €
    1 * 1 Pack.
       725,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-5739
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact
  • 613-5738
    3+1-pack, incl. three single channel pipettes (0,5 - 10 µl, 20 - 200 µl, 100 - 1000 µl), one 8-channel pipette (30 - 300 µl), Optifit Tip tray boxes (1× 0,1 - 10 µl, 1× 0,5 - 200 µl, 1× 5 - 350 µl, 1× 10 - 1000 µl) and a linear stand
    3+1-pack, incl. three single channel pipettes (0,5 - 10 µl, 20 - 200 µl, 100 - 1000 µl), one 8-channel pipette (30 - 300 µl), Optifit Tip tray boxes (1× 0,1 - 10 µl, 1× 0,5 - 200 µl, 1× 5 - 350 µl, 1× 10 - 1000 µl) and a linear stand
    Log in for availability
    Log in for availability
    1 * 1 Pack.
    1 * 1 Pack. - 1 310,00 €
    1 310,00 €
    1 * 1 Pack.
       1 310,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-5738
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact
  • 613-5734
    4-pack 20, incl. four single channel pipettes (0,5 - 10 µl, 2 - 20 µl, 20 - 200 µl, 100 - 1000 µl), Optifit Tip tray boxes (1× 0,1 - 10 µl, 2× 0,5 - 200 µl, 1× 10 - 1000 µl) and a linear stand
    4-pack 20, incl. four single channel pipettes (0,5 - 10 µl, 2 - 20 µl, 20 - 200 µl, 100 - 1000 µl), Optifit Tip tray boxes (1× 0,1 - 10 µl, 2× 0,5 - 200 µl, 1× 10 - 1000 µl) and a linear stand
    Log in for availability
    Log in for availability
    1 * 1 Pack.
    1 * 1 Pack. - 948,00 €
    948,00 €
    1 * 1 Pack.
       948,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-5734
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact
  • 613-5735
    4-pack 100, incl. four single channel pipettes (0,5 - 10 µl, 10 - 100 µl, 20 - 200 µl, 100 - 1000 µl), Optifit Tip tray boxes (1× 0,1 - 10 µl, 2× 0,5 - 200 µl, 1× 10 - 1000 µl) and a linear stand
    4-pack 100, incl. four single channel pipettes (0,5 - 10 µl, 10 - 100 µl, 20 - 200 µl, 100 - 1000 µl), Optifit Tip tray boxes (1× 0,1 - 10 µl, 2× 0,5 - 200 µl, 1× 10 - 1000 µl) and a linear stand
    Log in for availability
    Log in for availability
    1 * 1 Pack.
    1 * 1 Pack. - 948,00 €
    948,00 €
    1 * 1 Pack.
       948,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-5735
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact
  • 613-5736
    5-pack 10, incl. five single channel pipettes (0,5 - 10 µl, 10 - 100 µl, 20 - 200 µl, 100 - 1000 µl, 500 - 5000 µl), Optifit Tip tray boxes (1× 0,1 - 10 µl, 1× 0,5 - 200 µl, 1× 10 - 1000 µl, 1× 100 - 5000 µl) and a linear stand
    5-pack 10, incl. five single channel pipettes (0,5 - 10 µl, 10 - 100 µl, 20 - 200 µl, 100 - 1000 µl, 500 - 5000 µl), Optifit Tip tray boxes (1× 0,1 - 10 µl, 1× 0,5 - 200 µl, 1× 10 - 1000 µl, 1× 100 - 5000 µl) and a linear stand
    Log in for availability
    Log in for availability
    1 * 1 Pack.
    1 * 1 Pack. - 1 170,00 €
    1 170,00 €
    1 * 1 Pack.
       1 170,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-5736
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact
  • 613-5737
    5-pack 20, incl. five single channel pipettes (2 - 20 µl, 10 - 100 µl, 20 - 200 µl, 100 - 1000 µl, 500 - 5000 µl), Optifit Tip tray boxes (2× 0,5 - 200 µl, 1× 10 - 1000 µl, 1× 100 - 5000 µl) and a linear stand
    5-pack 20, incl. five single channel pipettes (2 - 20 µl, 10 - 100 µl, 20 - 200 µl, 100 - 1000 µl, 500 - 5000 µl), Optifit Tip tray boxes (2× 0,5 - 200 µl, 1× 10 - 1000 µl, 1× 100 - 5000 µl) and a linear stand
    Log in for availability
    Log in for availability
    1 * 1 Pack.
    1 * 1 Pack. - 1 170,00 €
    1 170,00 €
    1 * 1 Pack.
       1 170,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-5737
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact