Labels for BBP3x printers, type B-526
Supplier: Brady
Jump to:
About this item
Type B-526 phosphorescent polyester tape is designed for directional and evacuation marking. It is used for glow-in-the-dark identification up to 10 hours.
3 Options Available Below
Product Details & Documents
Type B-526 phosphorescent polyester tape is designed for directional and evacuation marking. It is used for glow-in-the-dark identification up to 10 hours.
Product Family Options
Product Information
- ModelMaterialAdhesivePrinting methodColourPrinter modelDimensionsPackedAvailabilityPrice
- BRDY117928Glow-in-the-dark label printer tapeGlow-in-the-dark label printer tapeB30C-1125-526PolyesterPermanent acrylicThermal transferGreenBBP3129 mm×15,2 m1 roll/boxLog in for availability1 * 1 Rolle279,00 €
- BRDY117926Glow-in-the-dark label printer tapeGlow-in-the-dark label printer tapeB30C-2250-526PolyesterPermanent acrylicThermal transferGreenBBP3157 mm×15,2 m1 roll/boxLog in for availability1 * 1 Rolle446,00 €
- BRDY117924Glow-in-the-dark label printer tapeGlow-in-the-dark label printer tapeB30C-4000-526PolyesterPermanent acrylicThermal transferGreenBBP31101 mm×15,2 m1 roll/boxLog in for availability1 * 1 Rolle696,00 €