Capillary columns, Rtx®-440 (fused silica)
: Restek
These general purpose capillary columns with unique selectivity for pesticides, PAHs, or other semivolatiles are ideal for low/trace-level analyses.
- Midpolarity proprietary Crossbond phase
- Low-bleed, high-resolution columns with unique selectivity
- Wide temperature range from 20 to 340 °C
Frequently Bought Together