Methane cylinder kit for GC
Catalog # REST20197
Supplier: Restek
Specifications
- Description:Methane cylinder kit
- Pk:1
Specifications
About this item
Set the column flow rate by injection the methane and optimising linear velocity is preferred method for establishing reproducible retention times.
Delivery: Kit includes a Scotty 14 cylinder containing 1% methane in helium, a MINICYL regulator, a syringe adaptor and a package of twenty-five septa for the adapter.