
Sulphuric acid ≥96%, SLSI for the electronics industry
Catalog # 50139211.
Supplier: BASF ELECTRONIC CHEMICALS
CAS Number:
Specifications
- Pack type:Plastic drum
- Pk:340 kg
- Seq:4811
Specifications
About this item
:
:98.08 g/mol
Merck Index:14,08974
Boiling Pt:~335 °C (1013 hPa)
Melting Pt:3 °C
Density:1,84 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-639-5
UN:1830
ADR:8,II
REACH:01-2119458838-20
:GHS05
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589