Spill Kits, in See-Thru bag
: NEW PIG
Compact, easy-to-inventory bag kit with zipper is perfect for stocking a fleet.
- Compact kit with zipper closure and carry handle is ideal for outfitting a vehicle fleet
- PIG socks stop spreading spills; PIG mat absorbs quickly
- Temporary disposal bags help make cleanup easier
- Bold Spill Kit lettering for quick identification
The transparent bag lets you check supply levels at a glance whilst keeping contents clean and dry.
: To ensure effectiveness and your safety, we recommend that you conduct compatibility and absorption testing of your chemicals with PIG® HAZ-MAT products prior to purchase.
Frequently Bought Together