Order Entry
Austria
ContactUsLinkComponent
Creatinine colorimetric detection kit
Creatinine colorimetric detection kit
Catalog # ENZOADI907030A
Supplier:  ENZO LIFE SCIENCES
undefined
Creatinine colorimetric detection kit
Catalog # ENZOADI907030A
Supplier:  ENZO LIFE SCIENCES

Specifications

  • Description:
    Creatinine colorimetric detection kit, includes microtiter plate, standard and detection reagent
  • Seq:
    02
  • Storage temperature:
    4 °C
  • Sensitivity:
    0,042 mg/dl (range 0,31 - 20 mg/dl)
  • Pk:
    1 KIT
  • For:
    Quantitative determination of urinary creatinine from any species

Specifications

About this item

The creatinine colorimetric detection kit utilises a single step liquid detection reagent that is safer and less time consuming than other assay methods.

  • Easy-to-use one step creatinine detection
  • Fast homogeneous mix-and-ready assay with no wash steps, results in just 30 minutes
  • Accurate and calibrated to NIST creatinine standard

This kit is calibrated against the NIST standard and offers reproducible results with less than 6% inter- and intra-assay variation.