HPLC columns, Pinnacle II PAH
: Restek
Pinnacle II PAH columns are developed specifically for challenging analyses of polycyclic aromatic hydrocarbons (PAHs).
- Reliable and cost-effective
- Greater control over quality and reproducibility
- Stationary phase incorporates a proprietary C18 bonding that enables unique shape selectivity
pH Range: 2,5 to 8
- Catalog No:
- REST9219452
- REST921950212
- REST921950210
- REST9219452-700
- REST9219453-700
- REST921941E
- REST9219475
- REST9219422
- REST9219475-700
- REST9219422-700
- REST9219423-700
- REST921947E
- REST9219425
- REST9219425-700
- REST921942E
- REST9219412-700
- REST9219472
- REST9219413-700
- REST9219472-700
- REST9219415
- REST9219415-700
- REST9219473-700
- REST9219455
- REST9219455-700
- REST921945E
- REST9219462
- REST9219462-700
- REST9219465
- REST9219465-700
- REST9219412
- Endcapped:
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- pH range:
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- Temperature limit:
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
Frequently Bought Together