Extraction cell caps and replacement parts
Supplier: Restek
Certificates
About this item
Caps features a smooth inner surfaces for easier cleaning. Suitable for for ASE systems.
Delivery: Caps include frit, PEEK washer, PTFE O-ring, threaded insert and snap ring.
Frequently Bought Together