About this item
This antibody recognizes a polypeptide which is identified as insulin, a 51-amino acid polypeptide composed of A and B chains connected through the C-peptide. Proinsulin, which has very little biological activity, is cleaved by proteases within its cell of origin into the insulin molecule and the C-terminal basic residue. Insulin enhances membrane transport of glucose, amino acids, and certain ions. It also promotes glycogen storage, formation of triglycerides, and synthesis of proteins and nucleic acids. Deficiency of insulin results in diabetes mellitus. The main storage site for insulin is the pancreatic islets. Antibodies to insulin are important as beta-cell and insulinoma marker.
Type: Primary
Antigen: INS
Clonality: Monoclonal
Clone: E2-E3 2D11-H5
Conjugation: CF647
Epitope:
Host: Mouse
Isotype: IgGs
Reactivity: Cow, Human, Mouse, Pig, Rabbit
Specifications
- Catalog No:
- BNC471226-100
- BNC471226-500
- Antigen symbol:
- INS
- INS
- Antigen name:
- Insulin
- Insulin
- Antigen synonyms:
- MODY10|ILPR|IDDM2|Proinsulin|IRDN
- MODY10|ILPR|IDDM2|Proinsulin|IRDN
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 647
- CF® 647
- Clone:
- Clone E2-E3 2D11-H5
- Clone E2-E3 2D11-H5
- Reactivity:
- Pig, Rabbit, Human, Bovine, Mouse
- Pig, Rabbit, Human, Bovine, Mouse
- Host:
- Mouse
- Mouse
- Gene ID:
- 272259
- 272259
- Isotype:
- IgGs
- IgGs
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide