Order Entry
Austria
ContactUsLinkComponent
Anti-INS Mouse Monoclonal Antibody (CF594) [clone: E2-E3 2D11-H5]
Anti-INS Mouse Monoclonal Antibody (CF594) [clone: E2-E3 2D11-H5]
Supplier:  Biotium
Certificates
Anti-INS Mouse Monoclonal Antibody (CF594) [clone: E2-E3 2D11-H5]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC941226-500
  • BNC941226-100
  • Antigen symbol:
  • INS
  • INS
  • Antigen name:
  • Insulin
  • Insulin
  • Antigen synonyms:
  • MODY10|ILPR|IDDM2|Proinsulin|IRDN
  • MODY10|ILPR|IDDM2|Proinsulin|IRDN
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 594
  • CF® 594
  • Clone:
  • Clone E2-E3 2D11-H5
  • Clone E2-E3 2D11-H5
  • Reactivity:
  • Pig, Rabbit, Human, Bovine, Mouse
  • Pig, Rabbit, Human, Bovine, Mouse
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 272259
  • 272259
  • Isotype:
  • IgGs
  • IgGs
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together