Anti-BRD4 Rabbit Polyclonal Antibody
ABGEAP8051A
: Abgent
- Antibody type:Primary
- Antigen symbol:BRD4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Epitope:N-Terminal (1–100 aa)
- Size:100 µg
- Pk:400 µl
Type: Primary
Antigen: BRD4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal (1–100 aa)
Host: Rabbit
Isotype:
Reactivity: Human