About this item
Recognizes a 47-55 kDa-tumor suppressor protein, identified as Wilm's Tumor (WT1) protein. The antibody reacts with all isoforms of the full-length WT1 and also identifies WT1 lacking exon 2-encoded amino acids, frequently found in subsets of sporadic Wilm s tumors.WT1, a sporadic and familial pediatric kidney tumor, is genetically heterogeneous. Wilm s tumor is associated with mutations of WT1, a zinc-finger transcription factor that is essential for the development of the metanephric kidney and the urogenital system. The WT1 gene is normally expressed in fetal kidney and mesothelium, and its expression has been suggested as a marker for Wilm s tumor and mesothelioma. WT1 protein has been identified in proliferative mesothelial cells, malignant mesothelioma, ovarian carcinoma, gonadoblastoma, nephroblastoma, and desmoplastic small round cell tumor. Lung adenocarcinomas rarely stain positive with this antibody. WT1 protein expression in mesothelial cells has become a reliable marker for the diagnosis of mesotheliomas.
- Purified
Type: Primary
Antigen: WT1
Clonality: Monoclonal
Clone: WT1/857 6F-H2
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- BTIUBNUB1138-500
- BTIUBNUB1138-100
- Antigen symbol:
- WT1
- WT1
- Antigen synonyms:
- WAGR|FWT1|AWT1|NPHS4|WT1|GUD|Wilms'tumor 1
- WAGR|FWT1|AWT1|NPHS4|WT1|GUD|Wilms'tumor 1
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- Clone WT1/857 6F-H2
- Clone WT1/857 6F-H2
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Mouse
- Mouse
- Gene ID:
- 234569
- 234569
- Isotype:
- IgG
- IgG
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,2 mg/ml
- 0,2 mg/ml
- Storage buffer:
- PBS/0,05% BSA/0,05% azide
- PBS/0,05% BSA/0,05% azide