Anti-PECAM1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # NOVUNBP1-71663H
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Platelet/endothelial cell adhesion molecule 1
- Antigen symbol:PECAM1
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Rabbit
- Reactivity:Human,Mouse
- Western blot:Yes
- Gene ID:5175
- Antigen synonyms:endoCAM|PECAM-1|GPIIA'|CD31|CD31/EndoCAM|PECA1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: PECAM1
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse