Order Entry
Austria
ContactUsLinkComponent
Anti-PECAM1 Rabbit Polyclonal Antibody (DyLight® 650)
Anti-PECAM1 Rabbit Polyclonal Antibody (DyLight® 650)
Catalog # NOVUNBP1-71663C
Supplier:  Novus Biologicals
Anti-PECAM1 Rabbit Polyclonal Antibody (DyLight® 650)
Catalog # NOVUNBP1-71663C
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Platelet/endothelial cell adhesion molecule 1
  • Antigen symbol:
    PECAM1
  • Clonality:
    Polyclonal
  • Conjugation:
    DyLight® 650
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Gene ID:
    5175
  • Antigen synonyms:
    endoCAM|PECAM-1|GPIIA'|CD31|CD31/EndoCAM|PECA1
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 μL
  • Pk:
    0,1 mL

Specifications

About this item

Type: Primary
Antigen: PECAM1
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse