Order Entry
Austria
ContactUsLinkComponent
 
Anti-Neural Stem Cell Marker Antibody
  NOVUNBP1-42826
undefined
Anti-Neural Stem Cell Marker Antibody
  NOVUNBP1-42826

 

  • Antigen name:
    Neural Stem Cell Marker
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Size:
    5 Vials
  • Pk:
    5 Vial

 

 

Neural Stem Cell Marker Antibody Pack. Tested Applications: Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry-Frozen, Immunohistochemistry-Paraffin. Tested Reactivity: Human, Mouse, Rat, Bovine, Canine, Chick

Type:
Antigen: Neural Stem Cell Marker
Clonality:
Clone:
Conjugation:
Epitope:
Host:
Isotype:
Reactivity: Human