Anti-PCSK9 Goat Polyclonal Antibody
Catalog # NOVUNB300-959
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Proprotein convertase subtilisin/kexin type 9
- Antigen symbol:PCSK9
- Clonality:Polyclonal
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Antigen synonyms:PCSK9|Neural apoptosis-regulated convertase 1|NARC-1|NARC1|Proprotein convertase subtilisin/kexin type 9|Subtilisin/kexin-like protease PC9|Proprotein convertase PC9|PSEC0052
- Size:100 μg
- Pk:0,1 mg
Specifications
About this item
Goat Polyclonal PCSK9 Antibody. Tested Applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA. Tested Reactivity: Human.
Recommended Dilutions: Immunohistochemistry (Parafin): 2-6 ?g/ml
Type: Primary
Antigen: PCSK9
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human