Anti-MAPK Rabbit Polyclonal Antibody
NOVUNB500-138
- Antibody type:Primary
- Antigen name:p38 MAPK
- Antigen symbol:MAPK
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Antigen synonyms:MAX-interacting protein 2|MAP kinase MXI2|CSAID-binding protein|MAPK 14|Stress-activated protein kin|CSBP|MAP kinase p38 alpha|Mitogen-activated protein kinase 14|MAP kinase 14|Mitogen-activated protein kinase p38 alpha|Cytokine suppressive anti-inflammatory drug-binding protein
- Size:100 μL
- Pk:0,1 mL
Rabbit Polyclonal p38 [p Tyr182] Antibody. Tested Applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen, Immunohistochemistry-Paraffin. Tested Reactivity: Human.
Type: Primary
Antigen: MAPK
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human