Order Entry
Austria
ContactUsLinkComponent
Anti-B3GAT1 Mouse Monoclonal Antibody (Purified) [clone: NK/804]
Anti-B3GAT1 Mouse Monoclonal Antibody (Purified) [clone: NK/804]
Supplier:  Biotium
Certificates
Anti-B3GAT1 Mouse Monoclonal Antibody (Purified) [clone: NK/804]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNUB0804-100
  • BNUB0804-500
  • Antigen symbol:
  • B3GAT1
  • B3GAT1
  • Antigen name:
  • CD57 / B3GAT1
  • CD57 / B3GAT1
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • Clone NK/804
  • Clone NK/804
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 381050
  • 381050
  • Isotype:
  • IgM kappa
  • IgM kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,2 mg/ml
  • 0,2 mg/ml
  • Storage buffer:
  • PBS/0,05% BSA/0,05% azide
  • PBS/0,05% BSA/0,05% azide

Specifications

Frequently Bought Together