Order Entry
Austria
ContactUsLinkComponent
Anti-INS Mouse Monoclonal Antibody (Purified, BSA-free) [clone: IRDN/794]
Anti-INS Mouse Monoclonal Antibody (Purified, BSA-free) [clone: IRDN/794]
Catalog # BNUM0794-50
Supplier:  Biotium
Anti-INS Mouse Monoclonal Antibody (Purified, BSA-free) [clone: IRDN/794]
Catalog # BNUM0794-50
Supplier:  Biotium

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Insulin
  • Antigen symbol:
    INS
  • Clonality:
    Monoclonal
  • Clone:
    Clone IRDN/794
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Pig,
    Human,
    Bovine,
    Rat,
    Mouse
  • Gene ID:
    272259
  • Antigen synonyms:
    MODY10|ILPR|IDDM2|Proinsulin|IRDN
  • Storage buffer:
    PBS/0,05% azide
  • Concentration:
    1 mg/mL
  • Purification:
    BSA-free
  • Size:
    50 μL
  • Pk:
    50 µl

Specifications

About this item

Recognizes a polypeptide which is identified as insulin, a 51-amino acid polypeptide composed of A and B chains connected through the C-peptide. Proinsulin, which has very little biological activity, is cleaved by proteases within its cell of origin into the insulin molecule and the C-terminal basic residue. Insulin enhances membrane transport of glucose, amino acids, and certain ions. It also promotes glycogen storage, formation of triglycerides, and synthesis of proteins and nucleic acids. Deficiency of insulin results in diabetes mellitus. The main storage site for insulin is the pancreatic islets. Antibodies to insulin are important as beta-cell and insulinoma marker.

Type: Primary
Antigen: INS
Clonality: Monoclonal
Clone: IRDN/794
Conjugation: Purified, BSA-free
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Cow, Human, Mouse, Pig, Rat