Order Entry
Austria
ContactUsLinkComponent
 
Anti-LCK Rabbit Polyclonal Antibody
  BOSSBS-2649R
 :  Bioss
undefined
Anti-LCK Rabbit Polyclonal Antibody
  BOSSBS-2649R
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    Lymphocyte-specific Protein tyrosine kinase
  • Antigen symbol:
    LCK
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Gene ID:
    3932
  • Antigen synonyms:
    p56lck|LSK|tyrosine-protein kinase Lck|YT16|leukocyte C-terminal Src kinase|T-lymphocyte specific protein tyrosine kinase p56lck|p56(LSTRA) protein-tyrosine kinase|RP4-675E8.4|t cell-specific protein-tyrosine kinase|lymphocyte cell-specific protein-tyrosine kinase|proto-oncogene tyrosine-protein kinase LCK|pp58lck
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
  • Storage temperature:
    Store at –20 °C for 12 months
  • Concentration:
    1 ug/ul
  • Shipping temperature:
    4 °C
  • Purification:
    Purified by Protein A
  • Size:
    100 μL
  • Pk:
    100 µl

 

 

Type: Primary
Antigen: LCK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat