Order Entry
Austria
ContactUsLinkComponent
Restriction enzymes, NheI, FastDigest™, fermentas
Restriction enzymes, NheI, FastDigest™, fermentas
Catalog # FD0974
CAS Number:  
undefined
Restriction enzymes, NheI, FastDigest™, fermentas
Catalog # FD0974
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Compatible buffer:
    FastDigest Green Buffer or FastDigest Buffer (colorless)
  • Overhang type:
    5' overhang
  • Description:
    NheI, FastDigest™
  • Enzyme name:
    FastDigest NheI
  • No. of reactions:
    100 (100 µl)

Specifications

About this item

Thermo Scientific™ FastDigest™ enzymes are an advanced line of restriction enzymes for rapid DNA digestion.

  • 100% activity of all FastDigest™ enzymes in one universal reaction buffer
  • Designed to complete digestion in 5 to 15 minutes
  • Direct loading on gels with FastDigest™ Green buffer
  • 100% buffer compatibility with downstream applications