Order Entry
Austria
ContactUsLinkComponent
 
Anti-CD8A Mouse Monoclonal Antibody (Purified) [clone: RIV11]
 :  Biotium
 
Anti-CD8A Mouse Monoclonal Antibody (Purified) [clone: RIV11]
 :  Biotium

 

  • Catalog No:
  • BNUB0333-500
  • BNUB0333-100
  • Antigen symbol:
  • CD8A
  • CD8A
  • Antigen name:
  • CD8
  • CD8
  • Antigen synonyms:
  • T8/Leu-2 T-lymphocyte differentiation antigen|LYT3|alpha polypeptide (p32)|T-cell surface glycoprotein CD8 alpha chain|CD8 antigen|MAL|Ly3
  • T8/Leu-2 T-lymphocyte differentiation antigen|LYT3|alpha polypeptide (p32)|T-cell surface glycoprotein CD8 alpha chain|CD8 antigen|MAL|Ly3
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • Clone RIV11
  • Clone RIV11
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 85258
  • 85258
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,2 mg/ml
  • 0,2 mg/ml
  • Storage buffer:
  • PBS/0,05% BSA/0,05% azide
  • PBS/0,05% BSA/0,05% azide

 

Frequently Bought Together