Order Entry
Austria
ContactUsLinkComponent
Nucleic acid extraction, MPure™ Viral/Pathogen nucleic acids extraction kit, MP Biomedicals
Nucleic acid extraction, MPure™ Viral/Pathogen nucleic acids extraction kit, MP Biomedicals
Catalog # ICNA117022130
Supplier:  MP Biomedicals
CAS Number:  
Nucleic acid extraction, MPure™ Viral/Pathogen nucleic acids extraction kit, MP Biomedicals
Catalog # ICNA117022130
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Method / Format:
    Magnetic beads
  • Prep size:
    Mini
  • Refine target molecule:
    Viral RNA
    Bacterial RNA
    Viral DNA
    Bacterial DNA
  • Sample type:
    Swabs
    Biofluids (serum, plasma, saliva)
  • Target molecule:
    Copurification of DNA & RNA
    DNA
    RNA
  • Description:
    MPure™ Viral/Pathogen Nucleic Acids Extraction Kit B, 48 tests
  • Seq:
    02
  • Description:
    MPure™ Viral/Pathogen Nucleic Acids Extraction Kit B, 48 tests

Specifications

About this item

MPure™ Viral/Pathogen Nucleic Acids Extraction Kit is used with the MPure-12™ Automated Nucleic Acid Purification System for purification of viral DNA/RNA and pathogen DNA from swab samples (cell-rich samples).