VWR® ARISTAR®, Cerium Standard Solutions for ICP-MS
Supplier: VWR Chemicals
Certificates
About this item
- Produced from high purity acids, water ASTM I 18 Megaohms
- Solution assayed by titration
- Final concentration verified against reference material from NIST
- Total maximum uncertainty: 0,5% (typical)
- Delivered with complete certificate of analysis
Certifications: Manufactured and tested in ISO 17034 / ISO 17025 accredited facilities
Limited Time Sale
Select items below are currently on sale
Frequently Bought Together