Orders LinkContactUsLinkComponent
Forceps, precision
 :   BOCHEM
Forceps, precision
 :  BOCHEM
Forceps, precision
Forceps, precision
These non serrated forceps are made of durable titanium.
  • High corrosion and thermal resistance
  • Lightweight, high strength
Titanium is resistant to diluted sulphuric acid, hydrochloric acid, solutions containing chloride, cold nitric acid as well as most organic acids and alkaline solutions, such as sodium hydroxide.
Product Family Options
  • Description
    Type
    Length
  • Precision forceps, extra sharp, straight (tower tip)
    Straight
    105 mm
    1 * 1 ST
    1 * 1 ST - 53,80 €
    53,80 €
    1 * 1 ST
       53,80 €
  • Precision forceps, extra sharp, curved
    Curved
    105 mm
    1 * 1 ST
    1 * 1 ST - 53,80 €
    53,80 €
    1 * 1 ST
       53,80 €
  • Precision forceps, straight, smooth tips, extra sharp
    Straight
    130 mm
    1 * 1 ST
    1 * 1 ST - 53,80 €
    53,80 €
    1 * 1 ST
       53,80 €