ContactUsLinkComponent
Disposable cuvettes
Disposable cuvettes
  VARI6610018700
 :  VARIAN
undefined
Disposable cuvettes
  VARI6610018700
 :  VARIAN
  • Path length:
    10 mm
  • Volume:
    1,5 ml
  • Window material:
    PS
  • Description:
    Rectangular cell
  • Pk:
    1
Increase flexibility and capabilities of spectrophotometers thanks to certified calibration and installation cells for optimum optical performance in any application.