Capillary columns, MXT®-200 (Siltek®-treated stainless steel)
Supplier: Restek
Certificates
About this item
MXT®-200 columns are general purpose columns for solvents, Freon® fluorocarbons, alcohols, ketones, silanes, and glycols.
- Midpolarity phase; Crossbond® trifluoropropylmethyl polysiloxane
- Temperature range up to 400 °C
- Equivalent to USP G6 phase
Coil diameter: 114,3 mm
Frequently Bought Together