Order Entry
Austria
ContactUsLinkComponent
 
DNA extraction, MPure™ Blood DNA extraction kits, MP Biomedicals
 
undefined
DNA extraction, MPure™ Blood DNA extraction kits, MP Biomedicals

 

  • Method / Format:
    Magnetic beads
  • Prep size:
    Mini
  • Refine target molecule:
    Total DNA
  • Sample type:
    Blood
  • Target molecule:
    DNA
  • Description:
    MPure™ Blood DNA Extraction Kit 200, 48 tests

 

Frequently Bought Together