ContactUsLinkComponent
VWR®, Drain Stoppers for VWB2 Water Baths
VWR®, Drain Stoppers for VWB2 Water Baths
Catalog # 462-0585
Supplier:  VWR Collection
VWR®, Drain Stoppers for VWB2 Water Baths
Catalog # 462-0585
Supplier:  VWR Collection
Specifications
  • For:
    VWB2 5, VWB2 12, VWB2 18, VWB2 26
  • Description:
    Drain stoppers, to prevent heat transfer beads entering the drain opening
  • Pk:
    5