Lysing matrix Z, MP Biomedicals
Supplier: MP Biomedicals
Certificates
About this item
Lysing Matrix Z is used primarily for lysis of animal and plant tissue, tougher stems, roots and seeds, and whole insects. Each impact-resistant tube contains yttria-stabilised zirconium oxide beads.
- Impact-resistant tube
- Ideal for cracking of very tough cell walls
- Lower shear rate minimises degradation
- Free from DNase and RNase
Matrix Z is an alternative to the very aggressive Matrix A because it's lower shear rate minimises degradation in applications such as RNA and protein isolation from very tough tissue.
Specifications
- Sample type:Plants; Animal Tissues; Wood; Seeds; Soil; Clay; Bacteria; Insects; Ancient and Dried Samples