VWR® OIML, Calibration Weights E1, with Calibration Certificate
Lieferant: VWR Collection
Zertifikate
Über diesen Artikel
Individual E1 OIML class weights are intended to ensure traceability between the national mass standards (with values derived from the International Prototype of the kilogram) and Class E2 and lower weights. They are also used with microbalances. Anti-magnetic, cobalt alloy weights are available from 1 to 500 mg. Packed in a plastic case, the wire weights are easy to store. Anti-magnetic, corrosion resistant, highly polished stainless steel knob weights are available from 1 to 200 g. Packed in a plastic case the weights are easy to store. Sets of weights are also available in either cobalt alloy or stainless steel with a choice of a plastic or wooden storage box.
- Identification number guarantees traceability for weights from 1 g upwards
- Laser engraving improves legibility of the markings
- Supplied with COFRAC certificate
Certifications: Conform to OILM R111 recommendations.
Packaging: Plastic cases have a vacuum sealed protective film to ensure that the case and calibration weight are protected during transportation.
Zugehörige Informationen
20352379_VWR_Calibration_Weights_E1_with_Calibration_Certificate_EN
Technisches Datenblatt
VWR® for Calibrations VWR® for Weighing
Produktliteratur
Frequently Bought Together